3,5-Dichloor-N-(1,1-dimethylprop-2-ynyl)benzamide ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P2374-1G P2374-5G
TCIAP2374-1GEA 19.8 EUR
TCIAP2374-1G TCIAP2374-5G
3,5-Dichloor-N-(1,1-dimethylprop-2-ynyl)benzamide ≥96.0% (by GC)
3,5-Dichloor-N-(1,1-dimethylprop-2-ynyl)benzamide
Formule: C₁₂H₁₁Cl₂NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00055346
CAS nummer: 23950-58-5
EINECS: 245-951-4
UN: 3077
ADR: 9,III
Merck Index: 14,07872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=787ea824ce9dd6c27e1fab19bf3393d14efec55b&vid=787ea824ce9dd6c27e1fab19bf3393d14efec55b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 155.0 to 159.0 deg-C