Phenyl anthranilate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P2400-1G P2400-5G
TCIAP2400-1GEA 38.5 EUR
TCIAP2400-1G TCIAP2400-5G
Phenyl anthranilate ≥98.0% (by GC)
Phenyl anthranilate
Formule: C₁₃H₁₁NO₂
MDL Nummer: MFCD00060603
CAS nummer: 10268-69-6
EINECS: 233-608-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR