Propyl vinyl ether ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Stabilisatie: Stabilised with potassium hydroxide
P2424-250ML P2424-25ML
TCIAP2424-250MLEA 82.5 EUR
TCIAP2424-250ML TCIAP2424-25ML
Propyl vinyl ether ≥97.0% (by GC) stabilised
Propyl vinyl ether
Formule: C₅H₁₀O
Kookpunt: 65°C
Vlampunt: –26°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00080707
CAS nummer: 764-47-6
EINECS: 212-123-9
UN: 3271
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %