2-Phenoxyethyl methacrylate ≥85.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with HQ + MEHQ

P2468-100G P2468-500G
TCIAP2468-100GEA 81.4 EUR
TCIAP2468-100G TCIAP2468-500G
2-Phenoxyethyl methacrylate ≥85.0% (by GC) stabilised
2-Phenoxyethyl methacrylate
Formule: C₁₂H₁₄O₃
Kookpunt: 260°C
Vlampunt: 100°C
MDL Nummer: MFCD00053685
CAS nummer: 10595-06-9
EINECS: 234-201-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3a77c1f0632555fb82de6477553bf61093dfc89&vid=c3a77c1f0632555fb82de6477553bf61093dfc89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR