Perfluorooctanonitrile ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P2483-1G
P2483-5G
TCIAP2483-1GEA
52.8
EUR
TCIAP2483-1G
TCIAP2483-5G
Perfluorooctanonitrile ≥96.0% (by GC)
Perfluorooctanonitrile
Formule:
C₈F₁₅N Kookpunt: 102°C |
MDL Nummer:
MFCD00039481 CAS nummer: 647-12-1 EINECS: 211-476-6 |
|
Specification Test Results
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...