Pelitinib (EKB-569) ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide, (E)-N-(4-(3-chloro-4-fluorophenylamino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide, EKB 569, Pelitinib, (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-dimethylamino-2-butenamide

P2529-25MG P2529-100MG
TCIAP2529-25MGEA 85.8 EUR
TCIAP2529-25MG TCIAP2529-100MG
Pelitinib (EKB-569) ≥97.0% (by HPLC)
Pelitinib (EKB-569)
Formule: C₂₄H₂₃ClFN₅O₂
Smeltpunt: 198°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD09837868
CAS nummer: 257933-82-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68e9d90e9d0c2f529201e14a9baa901434ece11f&vid=68e9d90e9d0c2f529201e14a9baa901434ece11f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR