Spautin-1 ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P2613-10MG P2613-50MG
TCIAP2613-10MGEA 128 EUR
TCIAP2613-10MG TCIAP2613-50MG
Spautin-1 ≥98.0% (by GC, titration analysis)
Spautin-1
Formule: C₁₅H₁₁F₂N₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD16962292
CAS nummer: 1262888-28-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR