n-Pentaan ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Pentane

P2621-100ML P2621-500ML
TCIAP2621-100MLEA 17.6 EUR
TCIAP2621-100ML TCIAP2621-500ML
n-Pentaan ≥97.0% (by GC)
n-Pentaan
Formule: H₃C(CH₂)₃CH₃
Kookpunt: 36°C
Vlampunt: –49°C
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
Merck Index: 14,07116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5addee6b378880884f6b289996b46de507c2880f&vid=5addee6b378880884f6b289996b46de507c2880f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR