2-Phenylpropan-2-yl Benzodithioate ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P2638-1G
P2638-5G
TCIAP2638-1GEA
169
EUR
TCIAP2638-1G
TCIAP2638-5G
2-Phenylpropan-2-yl Benzodithioate ≥98.0% (by HPLC)
2-Phenylpropan-2-yl Benzodithioate
Formule:
C₁₆H₁₆S₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD18252676 CAS nummer: 201611-77-0 |
Specification Test Results
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 35.0 to 39.0 deg-C |
Elemental analysis (Carbon) | 69.1 to 72.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...