2-Phenylethyl methacrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Phenethyl methacrylate, Methacrylic acid 2-Phenylethyl ester, Methacrylic acid Phenethyl ester

Stabilisatie: Stabilised with hydroquinone (HQ)

P2714-5G P2714-25G
TCIAP2714-5GEA 55 EUR
TCIAP2714-5G TCIAP2714-25G
2-Phenylethyl methacrylate ≥98.0% (by GC) stabilised
2-Phenylethyl methacrylate
Formule: C₁₂H₁₄O₂
MW: 190.24 g/mol
Kookpunt: 107°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00038232
CAS nummer: 3683-12-3
EINECS: 222-969-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR