4-(Pentafluorosulphanyl)phenyldiazonium tetrafluoroborate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P2723-1G P2723-5G
TCIAP2723-1GEA 152 EUR
TCIAP2723-1G TCIAP2723-5G
4-(Pentafluorosulphanyl)phenyldiazonium tetrafluoroborate ≥98.0% (by HPLC)
4-(Pentafluorosulphanyl)phenyldiazonium tetrafluoroborate
Formule: C₆H₄BF₉N₂S
Bewaar temperatuur: Koelkast
CAS nummer: 1605274-62-1
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR