Polyethylene glycol monododecyl ether n=∼23
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Polidocanol
P2737-25G P2737-500G
TCIAP2737-25GEA 17.6 EUR
TCIAP2737-25G TCIAP2737-500G
Polyethylene glycol monododecyl ether n=∼23
Polyethylene glycol monododecyl ether
Formule: HO(CH₂CH₂O)nC₁₂H₂₅
MDL Nummer: MFCD00080891
CAS nummer: 9002-92-0
EINECS: 500-002-6
UN: 3077
ADR: 9 III
Merck Index: 14,07558
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Freezing point 33.0 to 38.0 deg-C
The average value of n 21.0 to 25.0