Quinolinium dichromate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Q0058-10G Q0058-25G
TCIAQ0058-10GEA 66 EUR
TCIAQ0058-10G TCIAQ0058-25G
Quinolinium dichromate ≥97.0% (by titrimetric analysis)
Quinolinium dichromate
Formule: C₁₈H₁₆Cr₂N₂O₇
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00059108
CAS nummer: 56549-24-7
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR