ɑ-Quinquethiophene

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Q0079-100MG Q0079-500MG
TCIAQ0079-100MGEA 170 EUR
TCIAQ0079-100MG TCIAQ0079-500MG
ɑ-Quinquethiophene
alpha-Quinquethiophene
Formule: C₂₀H₁₂S₅
Smeltpunt: 253°C
CAS nummer: 5660-45-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Lambda max. 415.0 to 419.0 nm (CHCl3)
TOF-MS to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR