Riboflavin tetrabutyrate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Vitamin B2 Tetrabutyrate

R0055-25G R0055-5G
TCIAR0055-25GEA 157 EUR
TCIAR0055-25G TCIAR0055-5G
Riboflavin tetrabutyrate ≥98.0% (by HPLC)
Riboflavin tetrabutyrate
Formule: C₃₃H₄₄N₄O₁₀
MW: 656.73 g/mol
MDL Nummer: MFCD00059691
CAS nummer: 752-56-7
EINECS: 212-034-5
Merck Index: 14,08200

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 147.0 to 151.0 deg-C
Absorbance min. 0.270 (15 mg/L, EtOH, 443.0 to 447.0 nm)
abs-ratio (a/b) 2.47 to 2.77 (E270/E445)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR