Rolipram ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

R0110-50MG R0110-250MG
TCIAR0110-50MGEA 121 EUR
TCIAR0110-50MG TCIAR0110-250MG
Rolipram ≥98.0% (by HPLC)
Rolipram
Formule: C₁₆H₂₁NO₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00270906
CAS nummer: 61413-54-5
EINECS: 262-771-1
Merck Index: 14,08251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616c253617a86c2ead44de2b5aba318d969fa330&vid=616c253617a86c2ead44de2b5aba318d969fa330&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR