Ritodrine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
R0114-250MG
R0114-1G
TCIAR0114-250MGEA
23.1
EUR
TCIAR0114-250MG
TCIAR0114-1G
Ritodrine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ritodrine hydrochloride
Formule:
C₁₇H₂₁NO₃ Smeltpunt: 195°C |
MDL Nummer:
MFCD01657514 CAS nummer: 23239-51-2 EINECS: 245-514-8 Merck Index: 14,08237 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...