Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ru(OAc)2[(R)-binap]
R0166-200MG R0166-1G
TCIAR0166-200MGEA 39.6 EUR
TCIAR0166-200MG TCIAR0166-1G
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Formule: C₄₈H₃₈O₄P₂Ru
MW: 841.83 g/mol
MDL Nummer: MFCD09753020
CAS nummer: 325146-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e67c8f89b41908b0cd8be8e499e55df5b656be69&vid=e67c8f89b41908b0cd8be8e499e55df5b656be69&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Yellow to Amber to Dark purple powder to crystal
IR to pass test