(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

R0183-10MG R0183-50MG
TCIAR0183-10MGEA 83.6 EUR
TCIAR0183-10MG TCIAR0183-50MG
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one ≥98.0% (by HPLC)
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one
Formule: C₁₆H₂₁NO₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD03093861
CAS nummer: 85416-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2756ab06227bf3fdfd65dffe5b385d67443bc0ce&vid=2756ab06227bf3fdfd65dffe5b385d67443bc0ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 97.0 ee%
Elemental analysis (Nitrogen) 4.50 to 5.50 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR