Span® 83
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S0063-25G
S0063-500G
TCIAS0063-25GEA
17.6
EUR
TCIAS0063-25G
TCIAS0063-500G
Span® 83
Span® 83
Formule:
C₄₂H₇₆O₇ |
MDL Nummer:
MFCD00151568 CAS nummer: 8007-43-0 EINECS: 232-360-1 |
Specification Test Results
Appearance | Light yellow to Yellow clear liquid |
Saponification Value | 149.0 to 160.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...