Scopolamine methyl nitrate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Methscopolamine Nitrate

S0230-1G S0230-5G
TCIAS0230-1GEA 104 EUR
TCIAS0230-1G TCIAS0230-5G
Scopolamine methyl nitrate ≥98.0% (by HPLC, titration analysis)
Scopolamine methyl nitrate
Formule: C₁₈H₂₄N₂O₇
MW: 380.40 g/mol
MDL Nummer: MFCD00078239
CAS nummer: 6106-46-3
EINECS: 228-065-2
UN: 1544
ADR: 6.1 III
Merck Index: 14,08406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff2694ecc7814fe3b8728ca12e29eec3abcecc6&vid=8ff2694ecc7814fe3b8728ca12e29eec3abcecc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C
Specific rotation [a]20/D -23.0 to -27.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR