2-Salicylideneaminophenol ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

S0328-25G S0328-5G
TCIAS0328-25GEA 127 EUR
TCIAS0328-25G TCIAS0328-5G
2-Salicylideneaminophenol ≥98.0% (by titrimetric analysis)
2-Salicylideneaminophenol
Formule: C₁₃H₁₁NO₂
MDL Nummer: MFCD00020011
CAS nummer: 1761-56-4
EINECS: 217-166-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 185.0 to 189.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR