Glycidyl stearate ≥96.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Stearic Acid Glycidyl Ester
S0366-5G
S0366-25G
TCIAS0366-5GEA
115
EUR
TCIAS0366-5G
TCIAS0366-25G
Glycidyl stearate ≥96.0% (by titrimetric analysis)
Glycidyl stearate
Formule:
C₂₁H₄₀O₃ Kookpunt: 193°C |
MDL Nummer:
MFCD00059110 CAS nummer: 7460-84-6 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...