Methyl succinamate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Methyl 4-amino-4-oxobutanoate, Succinamic Acid Methyl Ester

S0479-25G S0479-5G
TCIAS0479-25GEA 108 EUR
TCIAS0479-25G TCIAS0479-5G
Methyl succinamate ≥98.0% (by GC)
Methyl succinamate
Formule: C₅H₉NO₃
Kookpunt: 118°C
MDL Nummer: MFCD08276419
CAS nummer: 53171-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fa33b8e09734244e3c9d6889e61ef0c2e4e5000&vid=0fa33b8e09734244e3c9d6889e61ef0c2e4e5000&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR