Silver sulphadiazine ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
S0595-25G
S0595-5G
TCIAS0595-25GEA
143
EUR
TCIAS0595-25G
TCIAS0595-5G
Silver sulphadiazine ≥98.0% (by HPLC, titration analysis)
Silver sulphadiazine
Formule:
C₁₀H₉AgN₄O₂S |
MDL Nummer:
MFCD00072101 CAS nummer: 22199-08-2 EINECS: 244-834-5 Merck Index: 14,08903 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...