Sucralose ≥98.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
S0839-25G S0839-5G S0839-100G
TCIAS0839-25GEA 34.1 EUR
TCIAS0839-25G TCIAS0839-5G TCIAS0839-100G
Sucralose ≥98.0% (by HPLC)
Sucralose
Formule: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
Smeltpunt: 130°C
MDL Nummer: MFCD03648615
CAS nummer: 56038-13-2
EINECS: 259-952-2
Merck Index: 14,08880
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +84.0 to +88.0 deg (C=1, H2O)