(3aR)-(+)-Sclareolide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

S0847-25G S0847-5G
TCIAS0847-25GEA 149 EUR
TCIAS0847-25G TCIAS0847-5G
(3aR)-(+)-Sclareolide ≥97.0% (by GC)
(3aR)-(+)-Sclareolide
Formule: C₁₆H₂₆O₂
MDL Nummer: MFCD00134168
CAS nummer: 564-20-5
EINECS: 209-269-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 122.0 to 126.0 deg-C
Specific rotation [a]20/D +43.0 to +47.0 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR