Sulphamide ≥96.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

S0890-25G S0890-250G
TCIAS0890-25GEA 25.3 EUR
TCIAS0890-25G TCIAS0890-250G
Sulphamide ≥96.0% (by total nitrogen basis)
Sulphamide
Formule: H₄N₂O₂S
MDL Nummer: MFCD00011606
CAS nummer: 7803-58-9
EINECS: 232-262-9
Merck Index: 14,08922

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR