Penicillin G potassium salt
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt
31749.03
31749.04
SERA31749.03EA
85
EUR
SERA31749.03
SERA31749.04
Penicillin G potassium salt
Penicillin G potassium salt
Formule:
C₁₆H₁₇KN₂O₄S Smeltpunt: 214…217 °C Dichtheid: 1,359 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00036193 CAS nummer: 113-98-4 EINECS: 204-038-0 Merck Index: 12,07225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...