Sclareol ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (1R,2R,4aS,8aS)-1-((R)-3-Hydroxy-3-methylpent-4-en-1-yl)-2,5,5,8a-tetramethyldecahydronaphthalen-2-ol

S0916-1G S0916-5G
TCIAS0916-1GEA 52.8 EUR
TCIAS0916-1G TCIAS0916-5G
Sclareol ≥96.0% (by GC)
Sclareol
Formule: C₂₀H₃₆O₂
Kookpunt: 189°C
MDL Nummer: MFCD00869558
CAS nummer: 515-03-7
EINECS: 208-194-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3d7fbae05f9c94ac2fa6e3f08e9c25e3311b04d&vid=d3d7fbae05f9c94ac2fa6e3f08e9c25e3311b04d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 99.0 to 103.0 deg-C
Specific rotation [a]20/D -5.0 to -3.0 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR