(S)-(-)-SEGPHOS® ≥99.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-(-)-4,4'-Bis(diphenylphosphino)-3,3'-bi(1,2-methylenedioxybenzene), (S)-(-)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole, (S)-(-)-SEGPHOS®

S0929-200MG S0929-1G
TCIAS0929-200MGEA 36.3 EUR
TCIAS0929-200MG TCIAS0929-1G
(S)-(-)-SEGPHOS® ≥99.0% (by HPLC)
(S)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole
Formule: C₃₈H₂₈O₄P₂
MW: 610.574 g/mol
Smeltpunt: 217°C
MDL Nummer: MFCD09753005
CAS nummer: 210169-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=812c8ce7e2f785db643ee397eb7150e3f2f3e4cf&vid=812c8ce7e2f785db643ee397eb7150e3f2f3e4cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Optical purity (LC) min. 99.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR