Safinamide ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionamide, (S)-2-((4-((3-Fluorobenzyl)oxy)benzyl)amino)propanamide

S0935-25MG S0935-100MG
TCIAS0935-25MGEA 30.8 EUR
TCIAS0935-25MG TCIAS0935-100MG
Safinamide ≥98.0% (by HPLC)
Safinamide
Formule: C₁₇H₁₉FN₂O₂
Smeltpunt: 212°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00897036
CAS nummer: 133865-89-1
Merck Index: 14,08315
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f713d73ce41fbdc1cd091b62d9917fdf42df8b6&vid=3f713d73ce41fbdc1cd091b62d9917fdf42df8b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.70 to 9.70 %
Specific rotation [a]20/D +15.0 to +19.0 deg (C=1, AcOH (95vol%))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR