Sodium Picosulphate monohydrate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S0936-100MG
S0936-1G
S0936-5G
TCIAS0936-100MGEA
31.9
EUR
TCIAS0936-100MG
TCIAS0936-1G
TCIAS0936-5G
Sodium Picosulphate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium Picosulphate monohydrate
Formule:
C₁₈H₁₃NNa₂O₈S₂·H₂O Smeltpunt: 275°C |
MDL Nummer:
MFCD00867640 CAS nummer: 1307301-38-7 EINECS: 233-120-9 Merck Index: 14,07406 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Absorbance (E1%1cm) | 110 to 130 (H2O,263.0 nm) |
Water | 3.0 to 4.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...