Sertindole ≥95.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-(2-(4-(5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl)piperidin-1-yl)ethyl)imidazolidin-2-one, 1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl]-1-piperidinyl]ethyl]-2-imidazolidinone
S0942-10MG
S0942-50MG
TCIAS0942-10MGEA
97.9
EUR
TCIAS0942-10MG
TCIAS0942-50MG
Sertindole ≥95.0% (by HPLC)
Sertindole
Formule:
C₂₄H₂₆ClFN₄O Smeltpunt: 155°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00867749 CAS nummer: 106516-24-9 Merck Index: 14,08466 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 12.00 to 13.00 % |