Stearyl Glycyrrhetinate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Glycyrrhetinic acid Octadecyl ester, Octadecyl Glycyrrhetinate, Glycyrrhetinic acid Stearyl ester

S0988-25G S0988-5G
TCIAS0988-25GEA 131 EUR
TCIAS0988-25G TCIAS0988-5G
Stearyl Glycyrrhetinate ≥98.0% (by HPLC)
Stearyl Glycyrrhetinate
Formule: C₄₈H₈₂O₄
MDL Nummer: MFCD00674112
CAS nummer: 13832-70-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR