Tripentylamine (mixture of branched chain isomers) ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0338-25ML T0338-500ML
TCIAT0338-25MLEA 25.3 EUR
TCIAT0338-25ML TCIAT0338-500ML
Tripentylamine (mixture of branched chain isomers) ≥97.0% (by titrimetric analysis)
Tripentylamine
Formule: C₁₅H₃₃N
Vlampunt: 91°C
MDL Nummer: MFCD00015270
CAS nummer: 621-77-2
EINECS: 210-705-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR