Trichloromelamine ≥95.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T0384-25G T0384-100G
TCIAT0384-25GEA 28.6 EUR
TCIAT0384-25G TCIAT0384-100G
Trichloromelamine ≥95.0% (by titrimetric analysis)
Trichloromelamine
Formule: C₃H₃Cl₃N₆
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006047
CAS nummer: 7673-09-8
EINECS: 231-648-4
UN: 1479
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdf82ac0225577a1007a9c6c1890adac350d17bd&vid=cdf82ac0225577a1007a9c6c1890adac350d17bd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (Iodometric Titration) min. 95.0 %