Triisodecyl phosphite (mixture of isomers) ≥95.0% (by NMR)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Phosphorous Acid Triisodecyl Ester
T0420-25ML
T0420-500ML
TCIAT0420-25MLEA
18.7
EUR
TCIAT0420-25ML
TCIAT0420-500ML
Triisodecyl phosphite (mixture of isomers) ≥95.0% (by NMR)
Triisodecyl phosphite (mixture of isomers)
Formule:
C₃₀H₆₃O₃P |
MDL Nummer:
MFCD00015059 CAS nummer: 25448-25-3 EINECS: 246-998-3 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | multi component mixture |
Purity (NMR) | min. 95.0 atom% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...