Phenylfluorone ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 9-Phenyl-2,3,7-trihydroxy-6-fluorone, 9-Phenyl-3-fluorone, 2,6,7-Trihydroxy-9-phenyl-3-isoxanthone

T0450-1G T0450-5G
TCIAT0450-1GEA 17.6 EUR
TCIAT0450-1G TCIAT0450-5G
Phenylfluorone ≥97.0% (by HPLC)
Phenylfluorone
Formule: C₁₉H₁₂O₅
MDL Nummer: MFCD00005048
CAS nummer: 975-17-7
EINECS: 213-550-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fef309f867358efb04597fcf05dcdba76cd898a6&vid=fef309f867358efb04597fcf05dcdba76cd898a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Absorbance min. 0.700 (7.5 mg/L, Methanol, 468.0 to 472.0 nm) (calcd.on dried substance)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR