Trimethylvinylammonium bromide ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Neurine Bromide
T0494-1G
T0494-10G
TCIAT0494-1GEA
37.4
EUR
TCIAT0494-1G
TCIAT0494-10G
Trimethylvinylammonium bromide ≥97.0% (by titrimetric analysis)
Trimethylvinylammonium bromide
Formule:
C₅H₁₂BrN |
MDL Nummer:
MFCD00050204 CAS nummer: 10603-92-6 EINECS: 234-227-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...