Trichloromethanesulphonyl chloride ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0611-25G T0611-5G
TCIAT0611-25GEA 281 EUR
TCIAT0611-25G TCIAT0611-5G
Trichloromethanesulphonyl chloride ≥95.0% (by titrimetric analysis)
Trichloromethanesulphonyl chloride
Formule: CCl₄O₂S
MW: 217.87 g/mol
MDL Nummer: MFCD00007452
CAS nummer: 2547-61-7
EINECS: 219-830-1
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR