1,2,4-Trivinylcyclohexane (mixture of isomers) ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2,4-Trivinylcyclohexane

T0899-25ML T0899-500ML
TCIAT0899-25MLEA 27.5 EUR
TCIAT0899-25ML TCIAT0899-500ML
1,2,4-Trivinylcyclohexane (mixture of isomers) ≥97.0% (by GC)
1,2,4-Trivinylcyclohexane (mixture of isomers)
Formule: C₁₂H₁₈
Kookpunt: 88°C
Vlampunt: 69°C
MDL Nummer: MFCD00001515
CAS nummer: 2855-27-8
EINECS: 220-668-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR