1,3,4,6-Tetrathiapentalene-2,5-dione ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T1132-1G T1132-25G T1132-5G
TCIAT1132-1GEA 77 EUR
TCIAT1132-1G TCIAT1132-25G TCIAT1132-5G
1,3,4,6-Tetrathiapentalene-2,5-dione ≥98.0% (by HPLC)
1,3,4,6-Tetrathiapentalene-2,5-dione
Formule: C₄O₂S₄
MDL Nummer: MFCD00010403
CAS nummer: 64394-45-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR