Tripentylamine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T1177-25ML
T1177-500ML
TCIAT1177-25MLEA
19.8
EUR
TCIAT1177-25ML
TCIAT1177-500ML
Tripentylamine ≥98.0% (by GC)
Tripentylamine
Formule:
C₁₅H₃₃N Kookpunt: 240°C Vlampunt: 81°C |
MDL Nummer:
MFCD00015270 CAS nummer: 621-77-2 EINECS: 210-705-7 UN: 2735 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...