Potassium-p-toluenethiosulphonate ≥98.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: p-Toluenethiosulphonic acid Potassium Salt
T1185-25G T1185-5G
TCIAT1185-25GEA 88 EUR
TCIAT1185-25G TCIAT1185-5G
Potassium-p-toluenethiosulphonate ≥98.0% (by HPLC)
Potassium-p-toluenethiosulphonate
Formule: C₇H₈O₂S₂
MW: 226.35 g/mol
Smeltpunt: 229°C
MDL Nummer: MFCD00003547
CAS nummer: 28519-50-8
EINECS: 249-065-9
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Drying loss 5.0 to 9.0 %