3a,4,7,7a-Tetrahydroindene ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

T1315-25ML T1315-500ML
TCIAT1315-25MLEA 56.1 EUR
TCIAT1315-25ML TCIAT1315-500ML
3a,4,7,7a-Tetrahydroindene ≥98.0% (by GC) stabilised
3a,4,7,7a-Tetrahydroindene
Formule: C₉H₁₂
Kookpunt: 160°C
Vlampunt: 38°C
MDL Nummer: MFCD00021227
CAS nummer: 3048-65-5
EINECS: 221-260-3
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR