5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

T1360-1G T1360-100MG
TCIAT1360-1GEA 50.6 EUR
TCIAT1360-1G TCIAT1360-100MG
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin ≥97.0% (by HPLC)
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin
Formule: C₄₈H₃₈N₄O₄
MW: 734.86 g/mol
MDL Nummer: MFCD00012072
CAS nummer: 22112-78-3

Order Now

Specification Test Results

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (HPLC) min. 97.0 area%
Lambda max. 420.0 to 424.0 nm (CHCl3)
Molar extinction coefficient min. 430000 (CHCl3, 420.0 to 424.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR