Methyl-2-(triphenylphosphoranylidene)acetate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T1363-25G T1363-100G
TCIAT1363-25GEA 64.9 EUR
TCIAT1363-25G TCIAT1363-100G
Methyl-2-(triphenylphosphoranylidene)acetate ≥98.0% (by HPLC, titration analysis)
Methyl-2-(triphenylphosphoranylidene)acetate
Formule: C₂₁H₁₉O₂P
MDL Nummer: MFCD00008455
CAS nummer: 2605-67-6
EINECS: 220-018-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf7cf4e13f7ecb27082d35eabddfea1299ed244a&vid=bf7cf4e13f7ecb27082d35eabddfea1299ed244a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ester Value) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR