Tribehenin ≥90.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Glycerol Tridocosanoate, Glyceryl tribehenate, Glyceryl behenate, 1,2,3-Tridocosanoyl Glycerol

T1391-1G T1391-5G
TCIAT1391-1GEA 59.4 EUR
TCIAT1391-1G TCIAT1391-5G
Tribehenin ≥90.0% (by titrimetric analysis)
Tribehenin
Formule: C₆₉H₁₃₄O₆
MW: 1059.83 g/mol
MDL Nummer: MFCD00056274
CAS nummer: 18641-57-1
EINECS: 242-471-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ester Value) min. 90.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR