m-Tolualdehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1448-1G T1448-10G
TCIAT1448-1GEA 37.4 EUR
TCIAT1448-1G TCIAT1448-10G
m-Tolualdehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
m-Tolualdehyde-2,4-dinitrophenylhydrazone
Formule: C₁₄H₁₂N₄O₄
MW: 300.27 g/mol
MDL Nummer: MFCD00191585
CAS nummer: 2880-05-9
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 212.0 to 215.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR